Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic acid, disodium salt, 99%, anhydrous

Catalog No. AC208745000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC208745000

Supplier: Thermo Scientific Chemicals 208745000

Only null left
Add to Cart

CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002790
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
Synonym disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color White
Loss on Drying 0.3% max. (120°C)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula NaO2CCH2CH2CO2Na
Quantity 500 g
Beilstein 02, 606
Merck Index 15, 8808
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 162.05
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Succinic acid, disodium salt, 0.99
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 205-778-7
RTECSNumber WM7751000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.