Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, SPEX CertiPrep™
SDP

Catalog No. S3301 Shop All Spex CertiPrep Products
Click to view available options
Quantity:
1 mL

Catalog No. S-3301

Supplier: SPEX CertiPrep S3301

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Succinic acid110-15-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 110-15-6, 7732-18-5
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix H2O
Quantity 1 mL
Chemical Name or Material Succinic Acid
Hazard Category Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.