Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinyl Chloride 95.0+%, TCI America™
SDP

Catalog No. S0110250G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.974
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl

Specifications

Color Yellow
Boiling Point 192°C
Quantity 250 g
UN Number 3265
Formula Weight 154.97
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Succinyl Chloride
RTECSNumber WN4900000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.