Learn More
Invitrogen™ Succinylated Lysine Polyclonal Antibody

Description
Positive test controls include: 293T, MCF7, C6, Mouse liver, Mouse heart.
Specifications
Specifications
| Antigen | Succinylated Lysine |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene Alias | Lysine; Succinyl; succinylated; succinylation |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide. |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.