Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUCLA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185860
Description
SUCLA2 Polyclonal specifically detects SUCLA2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SUCLA2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
A-BETA, ATP-specific succinyl-CoA synthetase subunit beta, beta subunit, EC 6.2.1, EC 6.2.1.5, mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta, MTDPS5, Renal carcinoma antigen NY-REN-39, succinate-CoA ligase, ADP-forming, beta subunit, succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SUCLA2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FGGIMRCDVIAQGIVMAVKDLEIKIPVVVRLQGTRVDDAKALIADSGLKILACDDLDEAARMVVKLSEIVTLAKQAHVDVKFQLP | |
0.1 mL | |
Stem Cell Markers | |
8803 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction