Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUCLA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576277 Supplier Invitrogen™ Supplier No. PA576277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ATP-specific succinyl-CoA synthetase functions in the citric acid cycle (TCA), coupling the hydrolysis of succinyl-CoA to the synthesis of ATP and thus represents the only step of substrate-level phosphorylation in the TCA. The beta subunit provides nucleotide specificity of the enzyme and binds the substrate succinate, while the binding sites for coenzyme A and phosphate are found in the alpha subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUCLA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SUCLA2
Gene Accession No. Q9P2R7, Q9Z2I9
Gene Alias 4930547K18Rik; A-BETA; A-SCS; ATP-specific succinyl-CoA synthetase subunit beta; ATP-specific succinyl-CoA synthetase, beta subunit; mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta; MTDPS5; renal carcinoma antigen NY-REN-39; RP11-528D24.2; SCSbetaA; SCS-betaA; succinate--CoA ligase (ADP-forming); Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial; succinate-CoA ligase ADP-forming beta subunit; succinate-CoA ligase beta subunit; succinate-CoA ligase, ADP-forming, beta subunit; succinate-Coenzyme A ligase, ADP-forming, beta subunit; succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial; succinyl-CoA synthetase beta-A chain; Sucla2
Gene Symbols SUCLA2
Host Species Rabbit
Immunogen Recombinant protein of human SUCLA2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20916, 361071, 8803
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.