Learn More
Invitrogen™ SUCLA2 Polyclonal Antibody

Description
Immunogen sequence: MAASMFYGRL VAVATLRNHR PRTAQRAAAQ VLGSSGLFNN HGLQVQQQQQ RNLSLHEYMS MELLQEAGVS VPKGYVAKSP DEAYAIAKKL GSKDVVIKAQ VLAGGRGKGT FESGLKGGVK IVFSPEEAKA VSSQMIGKKL FTKQTGEKGR ICNQVLVCER KYPRREYYFA ITMERSFQGP; Positive Samples: 293T, SW480, HeLa, Mouse heart, Mouse liver, Mouse brain, Rat liver; Cellular Location: Mitochondrion.
Specifications
Specifications
| Antigen | SUCLA2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.78 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SUCLA2 |
| Gene Accession No. | Q9P2R7, Q9Z2I9 |
| Gene Alias | 4930547K18Rik; A-BETA; A-SCS; ATP-specific succinyl-CoA synthetase subunit beta; ATP-specific succinyl-CoA synthetase, beta subunit; mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta; MTDPS5; renal carcinoma antigen NY-REN-39; RP11-528D24.2; SCSbetaA; SCS-betaA; succinate--CoA ligase (ADP-forming); Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial; succinate-CoA ligase ADP-forming beta subunit; succinate-CoA ligase beta subunit; succinate-CoA ligase, ADP-forming, beta subunit; succinate-Coenzyme A ligase, ADP-forming, beta subunit; succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial; succinyl-CoA synthetase beta-A chain; Sucla2 |
| Gene Symbols | SUCLA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.