Learn More
SUCLA2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305477AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog and Pig
ATP-specific succinyl-CoA synthetase functions in the citric acid cycle (TCA), coupling the hydrolysis of succinyl-CoA to the synthesis of ATP and thus represents the only step of substrate-level phosphorylation in the TCA (PubMed:15877282). The beta subunit provides nucleotide specificity of the enzyme and binds the substrate succinate, while the binding sites for coenzyme A and phosphate are found in the alpha subunit.Specifications
SUCLA2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9P2R7, Q9Z2I9 | |
Sucla2 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SUCLA2 | |
RP11-528D24.2, A-BETA, MTDPS5, SCS-betaA, ATP-specific succinyl-CoA synthetase subunit beta, ATP-specific succinyl-CoA synthetase, beta subunit, mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta, renal carcinoma antigen NY-REN-39, succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial, succinyl-CoA synthetase beta-A chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20916, 8803 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.