Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUCLA2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570534
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0534 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0534 Supplier Bethyl Laboratories, Inc Supplier No. A305478AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Pig and Golden hamster

ATP-specific succinyl-CoA synthetase functions in the citric acid cycle (TCA), coupling the hydrolysis of succinyl-CoA to the synthesis of ATP and thus represents the only step of substrate-level phosphorylation in the TCA (PubMed:15877282). The beta subunit provides nucleotide specificity of the enzyme and binds the substrate succinate, while the binding sites for coenzyme A and phosphate are found in the alpha subunit.

Specifications

Antigen SUCLA2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SUCLA2
Gene Accession No. Q9P2R7, Q9Z2I9
Gene Alias RP11-528D24.2, A-BETA, MTDPS5, SCS-betaA, ATP-specific succinyl-CoA synthetase subunit beta, ATP-specific succinyl-CoA synthetase, beta subunit, mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta, renal carcinoma antigen NY-REN-39, succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial, succinyl-CoA synthetase beta-A chain
Gene Symbols Sucla2
Host Species Rabbit
Immunogen Between 413 and 463
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20916, 8803
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.