Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUCLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SUCLA2 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8125
![]() ![]() |
Proteintech Group Inc
126271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SUCLA2 (succinate-CoA ligase, ADP-forming, beta subunit), also known as A-BETA, SCS-betaA or renal carcinoma antigen NY-REN-39, is a 463 amino acid mitochondrial matrix enzyme that belongs to the succinate/malate CoA ligase beta subunit family. Widely expressed, SUCLA2 dimerizes with the SCS alpha subunit to form SCS-A, an essential component of the tricarboxylic acid cycle. Defects in SUCLA2 may be involved in a group of autosomal recessive disorders known as Mitochondrial DNA depletion syndromes (MDSs) that are characterized by a decrease in mitochondrial DNA copy numbers in affected tissues.Specifications
SUCLA2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9P2R7, Q9Z2I9 | |
20916, 361071, 8803 | |
SUCLA2 Fusion Protein Ag3319 | |
Primary | |
-20°C | |
SUCLA2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A BETA, SCS betaA, SUCLA2 | |
Sucla2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title