Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUCLG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115983
Description
Antibody detects endogenous levels of total SUCLG1.
This gene encodes the alpha subunit of the heterodimeric enzyme succinate coenzyme A ligase. This enzyme is targeted to the mitochondria and catalyzes the conversion of succinyl CoA and ADP or GDP to succinate and ATP or GTP. Mutations in this gene are the cause of the metabolic disorder fatal infantile lactic acidosis and mitochondrial DNA depletion.
Specifications
| SUCLG1 | |
| Polyclonal | |
| Unconjugated | |
| SUCLG1 | |
| 1500000I01Rik; GALPHA; MTDPS9; SCS-alpha; Succinate--CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinate-CoA ligase alpha subunit; succinate-CoA ligase, alpha subunit; succinate-CoA ligase, GDP-forming alpha subunit; succinate-CoA ligase, GDP-forming, alpha subunit; succinyl-CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial; succinyl-CoA synthetase alpha subunit; succinyl-CoA synthetase subunit alpha; SUCLA1; Suclg1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114597, 56451, 8802 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P13086, P53597, Q9WUM5 | |
| SUCLG1 | |
| A synthesized peptide derived from human SUCLG1(Accession P53597), corresponding to amino acid residues T44-T94. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction