Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUCLG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557767 Supplier Invitrogen™ Supplier No. PA557767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASVIYVPPPF AAAAINEAIE AEIPLVVCIT EGIPQQDMVR VKHKLLRQEK TRLIGPNCPG VINPGECKIG IM Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

This gene encodes the alpha subunit of the heterodimeric enzyme succinate coenzyme A ligase. This enzyme is targeted to the mitochondria and catalyzes the conversion of succinyl CoA and ADP or GDP to succinate and ATP or GTP. Mutations in this gene are the cause of the metabolic disorder fatal infantile lactic acidosis and mitochondrial DNA depletion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUCLG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUCLG1
Gene Accession No. P53597
Gene Alias 1500000I01Rik; GALPHA; MTDPS9; SCS-alpha; Succinate--CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinate-CoA ligase alpha subunit; succinate-CoA ligase, alpha subunit; succinate-CoA ligase, GDP-forming alpha subunit; succinate-CoA ligase, GDP-forming, alpha subunit; succinyl-CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial; succinyl-CoA synthetase alpha subunit; succinyl-CoA synthetase subunit alpha; SUCLA1; Suclg1
Gene Symbols SUCLG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUCLG1. Recombinant protein control fragment (Product #RP-102734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.