Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUCLG2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297953JF549
Description
SUCLG2 Polyclonal specifically detects SUCLG2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SUCLG2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 6.2.1, EC 6.2.1.4, GBETA, GTP-specific succinyl-CoA synthetase subunit beta, mitochondrial, succinate-CoA ligase, GDP-forming, beta subunit, succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial, succinyl-CoA synthetase, beta-G chain | |
Produced in rabbits immunized with E. coli-derived Human SUCLG2 fragment. | |
0.1 mL | |
Stem Cell Markers | |
8801 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction