Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUCLG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561289 Supplier Invitrogen™ Supplier No. PA561289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGGIVNCAII ANGITKACRE LELKVPLVVR LEGTNVQEAQ KILNNSGLPI TSAIDLEDAA KKAVASVAKK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

This gene encodes a GTP-specific beta subunit of succinyl-CoA synthetase. Succinyl-CoA synthetase catalyzes the reversible reaction involving the formation of succinyl-CoA and succinate. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 5 and 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUCLG2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUCLG2
Gene Accession No. Q96I99
Gene Alias AF171077; AW556404; D6Wsu120e; GBETA; G-BETA; G-SCS; GTPSCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; hypothetical protein LOC537713; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinate-CoA ligase GDP-forming subunit beta; succinate-CoA ligase, GDP-forming, beta subunit; succinate-Coenzyme A ligase, GDP-forming, beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; Suclg2
Gene Symbols SUCLG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUCLG2. Recombinant protein control fragment (Product #RP-99908).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.