Learn More
Invitrogen™ SUCLG2 Polyclonal Antibody

Description
Immunogen sequence: SDNGVRVQRF FVADTANEAL EAAKRLNAKE IVLKAQILAG GRGKGVFNSG LKGGVHLTKD PNVVGQLAKQ MIGYNLATKQ TPKEGVKVNK VMVAEALDIS RETYLAILMD RSCNGPVLVG SPQGGVDIEE VAASNPELIF KEQIDIFEGI KDSQAQRMAE NLGFVGPLKS QAADQITKLY NLFLKIDATQ VEVNPFGETP EGQVVCFDAK INFDDNAEFR QKDIFAMDDK SENEPIENEA AKYDLKYIGL D; Positive Samples: HT-29, 293T, LO2, U-87MG, MCF7, Raji, Mouse spleen, Rat spleen; Cellular Location: Mitochondrion.
Specifications
Specifications
| Antigen | SUCLG2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.56 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SUCLG2 |
| Gene Accession No. | Q96I99, Q9Z2I8 |
| Gene Alias | AF171077; AW556404; D6Wsu120e; GBETA; G-BETA; G-SCS; GTPSCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; hypothetical protein LOC537713; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinate-CoA ligase GDP-forming subunit beta; succinate-CoA ligase, GDP-forming, beta subunit; succinate-Coenzyme A ligase, GDP-forming, beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; Suclg2 |
| Gene Symbols | SUCLG2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.