Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUCLG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598728
Description
SUCLG2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a GTP-specific beta subunit of succinyl-CoA synthetase. Succinyl-CoA synthetase catalyzes the reversible reaction involving the formation of succinyl-CoA and succinate. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 5 and 12.
Specifications
SUCLG2 | |
Polyclonal | |
Unconjugated | |
SUCLG2 | |
AF171077; AW556404; D6Wsu120e; GBETA; G-BETA; G-SCS; GTPSCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; hypothetical protein LOC537713; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinate-CoA ligase GDP-forming subunit beta; succinate-CoA ligase, GDP-forming, beta subunit; succinate-Coenzyme A ligase, GDP-forming, beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; Suclg2 | |
Rabbit | |
Protein G | |
RUO | |
8801 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.33 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96I99 | |
SUCLG2 | |
Recombinant Human Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial protein (161-292AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction