Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUCNR1/GPR91 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2823500.1ML
Description
SUCNR1/GPR91 Polyclonal specifically detects SUCNR1/GPR91 in Human samples. It is validated for Western Blot.Specifications
SUCNR1/GPR91 | |
Polyclonal | |
PBS, 2% sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1 | |
The immunogen is a synthetic peptide directed towards the middle region of Human SUCNR1 | |
0.1 mL | |
GPCR | |
56670 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction