Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUCO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561478 Supplier Invitrogen™ Supplier No. PA561478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KWCSVRVALY RQRSRTALSK GKDYLVLAQP PLLLPAESVD VSVLQPLSGE LENTNIEREA ETVVLGDLSS SMHQDDLVNH TVDAVELEPS HSQTLSQSLL L Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 66%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SUCO
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Suco
Gene Accession No. Q9UBS9
Gene Alias 4732491B11; AI848100; AU016931; C1orf9; CH1; Membrane protein CH1; OPT; osteopotentia; protein C1orf9 homolog; protein osteopotentia; Protein osteopotentia homolog; SLP1; Suco; SUN domain containing ossification factor; SUN domain-containing ossification factor; SUN-like protein 1
Gene Symbols Suco
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUCO. Recombinant protein control fragment (Product #RP-99779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.