Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucralfate, Fine Powder, USP, 30-38%, Spectrum™ Chemical
SDP

Catalog No. 18607066 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

Catalog No. 18-607-066

Supplier: Spectrum Chemical Manufacturing Corporation S17645KGBL

Only null left
Add to Cart
Add to Cart

S1764, 54182-58-0, C12H54Al16O75S8

Spectrum™ Chemical Sucralfate, Fine Powder, USP is an oral gastrointestinal medicine used in the treatment of duodenal ulcers. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54182-58-0

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 5 kg
Percent Purity 30 to 38%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.