Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucralose, NF, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607075
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail

S1416, 56038-13-2, C12H19Cl3O8

Spectrum™ Chemical Sucralose, NF is used as an artificial sweetener in phamaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56038-13-2
Molecular Formula C12H19Cl3O8
Molecular Weight (g/mol) 397.63
MDL Number MFCD03648615
InChI Key BAQAVOSOZGMPRM-QBMZZYIRSA-N
IUPAC Name (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
SMILES OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

Specifications

CAS Max % 1
Ignition Residue 0.007
Packaging Poly Pail
Quantity 2.5 kg
Specific Rotation +84.0° to +87.5°
Percent Purity 98 to 102%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.