Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose, ACS, MilliporeSigma™

Catalog No. MSX10751 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX10751 500 g
MSX10753 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MSX10751 Supplier MilliporeSigma Supplier No. SX10751

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color White
Linear Formula C12H22O11
Quantity 500 g
Formula Weight 342.3
Grade ACS
Physical Form Crystalline
Chemical Name or Material Sucrose, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.