Learn More
Sudan IV (Solvent Red 24), Reagents
Specifications
Specifications
| Chemical Name or Material | Sudan IV (Solvent Red 24) |
| CAS | 85-83-6 |
| Molecular Formula | C24H20N4O |
| Synonym | Solvent Red 24 |
| Molecular Weight (g/mol) | 380.4418 |
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| Color | Dark Brown |
| Quantity | 10 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.