Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) chloride, 99.995%, (trace metal basis)

Catalog No. AC193190500
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle

Catalog No. AC193190500

Supplier: Thermo Scientific Chemicals 193190500

Only null left
Add to Cart

CAS: 10025-82-8 | Cl3In | 221.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
Molecular Formula Cl3In
Molecular Weight (g/mol) 221.18

Specifications

Melting Point 586.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.995%
Linear Formula InCl3
Merck Index 15,4998
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene
Formula Weight 221.18
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 233-043-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.