Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUFU Monoclonal Antibody (1783CT536.263.29)
GREENER_CHOICE

Catalog No. PIMA537736
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537736 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537736 Supplier Invitrogen™ Supplier No. MA537736
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUFU Monoclonal Antibody for Western Blot, IHC (P)

The Hedgehog signaling pathway plays an important role in early human development. The pathway is a signaling cascade that plays a role in pattern formation and cellular proliferation during development. This gene encodes a negative regulator of the hedgehog signaling pathway. Defects in this gene are a cause of medulloblastoma. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUFU
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1783CT536.263.29
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SUFU
Gene Accession No. Q9UMX1, Q9Z0P7
Gene Alias OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; PRO1280; RP11-47A8.1; Su(fu); Sufu; SUFU negative regulator of hedgehog signaling; SUFUH; SUFUXL; suppressor of fused homolog; suppressor of fused homolog (Drosophila); UNQ650/PRO1280
Gene Symbols SUFU
Host Species Mouse
Immunogen recombinant protein of human SUFU.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24069, 51684
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.