Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUFU Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589230 Supplier Invitrogen™ Supplier No. PA589230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse testis, Rat kidney; Cellular Location: Cytoplasm, Nucleus.

The Hedgehog signaling pathway plays an important role in early human development. The pathway is a signaling cascade that plays a role in pattern formation and cellular proliferation during development. This gene encodes a negative regulator of the hedgehog signaling pathway. Defects in this gene are a cause of medulloblastoma. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUFU
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SUFU
Gene Accession No. Q9UMX1, Q9Z0P7
Gene Alias OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; PRO1280; RP11-47A8.1; Su(fu); Sufu; SUFU negative regulator of hedgehog signaling; SUFUH; SUFUXL; suppressor of fused homolog; suppressor of fused homolog (Drosophila); UNQ650/PRO1280
Gene Symbols SUFU
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24069, 361769, 51684
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.