Learn More
SuFu Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302335A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).Specifications
SuFu | |
Polyclonal | |
Unconjugated | |
SUFU | |
OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; suppressor of fused homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24069, 51684 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UMX1, Q9Z0P7 | |
Sufu | |
Between 434 and 484 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.