Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUG1 Antibody (25D5), Novus Biologicals™
SDP

Catalog No. NB100345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100345 Supplier Novus Biologicals Supplier No. NB100345
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

SUG1 Monoclonal specifically detects SUG1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Knockdown Validated.

Specifications

Antigen SUG1
Applications Western Blot, Immunoprecipitation, ChIP Assay, KnockDown
Classification Monoclonal
Clone 25D5
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Knockdown Validated
Gene Accession No. P62195
Gene Alias MSUG1 protein, p45, p45/SUG26S protease regulatory subunit 8, proteasome (prosome, macropain) 26S subunit, ATPase, 5, proteasome 26S ATPase subunit 5, Proteasome 26S subunit ATPase 5, Proteasome subunit p45, S8, SUG-1, SUG126S proteasome AAA-ATPase subunit RPT6, Tat-binding protein homolog 10, TBP10, thyroid receptor interactor 1, TRIP1Thyroid hormone receptor-interacting protein 1
Gene Symbols PSMC5
Host Species Mouse
Immunogen Recombinant protein encoding full-length SUG1 purified from E. coli.
Molecular Weight of Antigen 46 kDa
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5705
Test Specificity SUG1 (25D5).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.