Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570575 Supplier Invitrogen™ Supplier No. PA570575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 91%.

SUGCT (Succinyl-CoA:Glutarate-CoA Transferase) is a Protein Coding gene. Diseases associated with SUGCT include Glutaric Aciduria Iii and Noonan Syndrome 8. Gene Ontology (GO) annotations related to this gene include succinate-hydroxymethylglutarate CoA-transferase activity. An important paralog of this gene is AMACR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGCT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Sugct
Gene Accession No. Q7TNE1
Gene Alias 5033411D12Rik; C18H7orf10; C22orf6; C4H7orf10; C7orf10; D17907; dermal papilla derived protein 13; dermal papilla-derived protein 13; DERP13; GA3; hypothetical protein LOC619244; KIAA1186; KIAA1658; LOW QUALITY PROTEIN: succinate--hydroxymethylglutarate CoA-transferase; ORF19; RGD1308114; Russel-Silver syndrome candidate; succinate--hydroxymethylglutarate CoA-transferase; succinyl-CoA glutarate-CoA transferase; succinyl-CoA:glutarate-CoA transferase; succinylCoA:glutarate-CoA transferase; SUGCT; zgc:113390
Gene Symbols Sugct
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SUGCT (aa 247-296).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192136
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.