Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555384 Supplier Invitrogen™ Supplier No. PA555384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGVKIIKELA AVCDVFVENY VPGKLSAMGL GYEDIDEIAP HIIYCSITGY GQTGPISQRA GYDAVASAVS GLMHITGPEN GDPVRPGVAM TDLATGLYAY GAIMAGLIQK YKTGKGLFID CNLLS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 29%.

SUGCT (Succinyl-CoA:Glutarate-CoA Transferase) is a Protein Coding gene. Diseases associated with SUGCT include Glutaric Aciduria Iii and Noonan Syndrome 8. Gene Ontology (GO) annotations related to this gene include succinate-hydroxymethylglutarate CoA-transferase activity. An important paralog of this gene is AMACR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGCT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sugct
Gene Accession No. Q9HAC7
Gene Alias 5033411D12Rik; C18H7orf10; C22orf6; C4H7orf10; C7orf10; D17907; dermal papilla derived protein 13; dermal papilla-derived protein 13; DERP13; GA3; hypothetical protein LOC619244; KIAA1186; KIAA1658; LOW QUALITY PROTEIN: succinate--hydroxymethylglutarate CoA-transferase; ORF19; RGD1308114; Russel-Silver syndrome candidate; succinate--hydroxymethylglutarate CoA-transferase; succinyl-CoA glutarate-CoA transferase; succinyl-CoA:glutarate-CoA transferase; succinylCoA:glutarate-CoA transferase; SUGCT; zgc:113390
Gene Symbols Sugct
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUGCT. Recombinant protein control fragment (Product #RP-92800).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.