Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552130 Supplier Invitrogen™ Supplier No. PA552130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKANRWFGVA PPKSGKMNMN ILHQEELIAQ KKREIEAKME QKAKQNQVAS PQPPHPGEIT NAHNSSCISN KFANDGSFLQ QFLKLQKAQT STDAPTSAPS APPSTPT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.

SF4 is a member of the SURP family of splicing factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUGP1
Gene Accession No. Q68FU8, Q8CH02, Q8IWZ8
Gene Alias 5730496N02Rik; DKFZp434E2216; F23858; RBP; RNA-binding protein RBP; SF4; Splicing factor 4; SUGP1; SURP and G patch domain containing 1; SURP and G-patch domain containing 1; SURP and G-patch domain-containing protein 1
Gene Symbols SUGP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUGP1. Recombinant protein control fragment (Product #RP-89200).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290666, 57794, 70616
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.