Learn More
Invitrogen™ SUGP2 Polyclonal Antibody

Description
Immunogen sequence: DQLVKRVIEG SLSPKERTLL KEDPAYWFLS DENSLEYKYY KLKLAEMQRM SENLRGADQK PTSADCAVRA MLYSRAVRNL KKKLL Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 69%.
Specifications
Specifications
| Antigen | SUGP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SUGP2 |
| Gene Accession No. | Q8IX01 |
| Gene Alias | Arginine/serine-rich-splicing factor 14; KIAA0365; mKIAA0365; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; SFRS14; Splicing factor, arginine/serine-rich 14; Srsf14; SUGP2; SURP and G patch domain containing 2; SURP and G-patch domain containing 2; SURP and G-patch domain-containing protein 2 |
| Gene Symbols | SUGP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.