Learn More
Invitrogen™ SUGP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590422
Description
Immunogen sequence: EAVGLQDIAP SPAAFPNFED STLFGREYID HLKAWLVSSG CPLQVKKAEP EPMREEEKMI PPTKPEIQAK APSSLSDAVP QRADHRVVGT IDQLVKRVIE GSLSPKERTL LKEDPAYWFL SDENSLEYKY YKLKLAEMQR MSENLRGADQ KPTSADCAVR AMLYSRAVRN LKKKLLPWQR RGLLRAQGLR GWKARRATTG T; Positive Samples: U-251MG, 293T, LO2, Jurkat, HeLa, Mouse thymus; Cellular Location: Nucleus.
This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described. [provided by RefSeq].
Specifications
SUGP2 | |
Polyclonal | |
Unconjugated | |
SUGP2 | |
Arginine/serine-rich-splicing factor 14; KIAA0365; mKIAA0365; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; SFRS14; Splicing factor, arginine/serine-rich 14; Srsf14; SUGP2; SURP and G patch domain containing 2; SURP and G-patch domain containing 2; SURP and G-patch domain-containing protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10147, 234373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8CH09, Q8IX01 | |
SUGP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human SUGP2 (NP_0556992). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.