Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104049 Supplier Invitrogen™ Supplier No. PA5104049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRS14.

This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SUGP2
Gene Accession No. Q8IX01
Gene Alias Arginine/serine-rich-splicing factor 14; KIAA0365; mKIAA0365; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; SFRS14; Splicing factor, arginine/serine-rich 14; Srsf14; SUGP2; SURP and G patch domain containing 2; SURP and G-patch domain containing 2; SURP and G-patch domain-containing protein 2
Gene Symbols SUGP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SUGP2(Accession Q8IX01), corresponding to amino acid residues L282-I332.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10147
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.