Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGP2 Polyclonal Antibody

Catalog No. PIPA535979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535979 Supplier Invitrogen™ Supplier No. PA535979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa NE, Neuro 2A, C2C12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SUGP2
Gene Accession No. Q8CH09, Q8IX01
Gene Alias Arginine/serine-rich-splicing factor 14; KIAA0365; mKIAA0365; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; SFRS14; Splicing factor, arginine/serine-rich 14; Srsf14; SUGP2; SURP and G patch domain containing 2; SURP and G-patch domain containing 2; SURP and G-patch domain-containing protein 2
Gene Symbols SUGP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SFRS14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10147, 234373
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.