Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUGT1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149707AF532
Description
SUGT1 Polyclonal antibody specifically detects SUGT1 in Human, Mouse samples. It is validated for Western BlotSpecifications
SUGT1 | |
Polyclonal | |
Western Blot | |
Protein 40-6-3, putative 40-6-3 protein, Sgt1, SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae), SGT1B protein, suppressor of G2 allele of SKP1 homolog, suppressor of G2 allele of SKP1, S. cerevisiae, homolog of | |
The immunogen for this product maps to a region between residue 315 and 365 of human Suppressor of G2 Allele of SKP1 using the numbering given in entry NP_001124384.1 (GeneID 10910). | |
0.1 mL | |
Cell Cycle and Replication | |
10910 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction