Learn More
Invitrogen™ SUGT1 Monoclonal Antibody (OTI7D2)

Description
SUGT1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SUGT1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI7D2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SUGT1 |
| Gene Accession No. | Q9Y2Z0 |
| Gene Alias | 2410174K12Rik; 40-6-3; Aa1114; LOC101122608; MGC137150 protein; Protein 40-6-3; Protein SGT1 homolog; putative 40-6-3 protein; Sgt1; SGT1 homolog, MIS12 kinetochore complex assembly cochaperone; SGT1, suppressor of G2 allele of SKP1; SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae); SGT1B protein; SUGT1; suppressor of G2 allele of SKP1; suppressor of G2 allele of SKP1 homolog; suppressor of G2 allele of SKP1, S. cerevisiae, homolog of; suppressor of g2 allele of SKP1-like protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.