Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUGT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583695 Supplier Invitrogen™ Supplier No. PA583695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEKNEKLEGD AALNRLFQQI YSDGSDEVKR AMNKSFMESG GTVLSTNWSD VGKRKVEINP PDDMEWK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

This gene is homologous to the yeast gene SGT1, which encodes a protein involved in kinetochore function and required for the G1/S and G2/M transitions. Complementation studies suggest that the human protein has similar functions. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUGT1
Gene Accession No. Q9Y2Z0
Gene Alias 2410174K12Rik; 40-6-3; Aa1114; LOC101122608; MGC137150 protein; Protein 40-6-3; Protein SGT1 homolog; putative 40-6-3 protein; Sgt1; SGT1 homolog, MIS12 kinetochore complex assembly cochaperone; SGT1, suppressor of G2 allele of SKP1; SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae); SGT1B protein; SUGT1; suppressor of G2 allele of SKP1; suppressor of G2 allele of SKP1 homolog; suppressor of G2 allele of SKP1, S. cerevisiae, homolog of; suppressor of g2 allele of SKP1-like protein
Gene Symbols SUGT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUGT1. Recombinant protein control fragment (Product #RP-102952).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10910
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.