Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUHW1 Antibody (OTI2H3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274409AF350
Description
SUHW1 Monoclonal specifically detects SUHW1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
SUHW1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ZNF280A | |
Recombinant protein fragment corresponding to amino acids 1-333 of human SUHW1 (NP_542778) produced in E.coli. | |
0.1 mL | |
129025.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI2H3 | |
Western Blot, Flow Cytometry | |
C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction