Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SULF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550890
Description
The antibody detects endogenous levels of total SULF1 protein.
Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.
Specifications
SULF1 | |
Polyclonal | |
Unconjugated | |
SULF1 | |
AI467640; AW121680; extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; Kiaa1077; mKIAA1077; mSulf-1; RSulfFP1; Sulf1; SULF-1; sulfatase 1; sulfatase FP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23213 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IWU6 | |
SULF1 | |
Synthetic peptide corresponding to a region derived from internal residues of human sulfatase 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction