Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589445 Supplier Invitrogen™ Supplier No. PA589445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDLLTVRQST RNLYARGFHD KDKECSCRES GYRASRSQRK SQRQFLRNQG TPKYKPRFVH TRQTRSLSVE FEGEIYDINL EEEEELQVLQ PRNIAKRHDE GHKGPRDLQA SSGGNRGRML ADSSNAVGPP T Positive Samples: 293T, U-87MG; Cellular Location: Cell surface, Endoplasmic reticulum, Golgi apparatus, Golgi stack.

Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SULF1
Gene Accession No. Q8IWU6, Q8VI60
Gene Alias AI467640; AW121680; extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; Kiaa1077; mKIAA1077; mSulf-1; RSulfFP1; Sulf1; SULF-1; sulfatase 1; sulfatase FP
Gene Symbols SULF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 480-610 of human SULF1 (NP_0011216761).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171396, 23213
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.