Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113112
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113112 Supplier Invitrogen™ Supplier No. PA5113112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SULF1 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SULF1
Gene Accession No. Q8IWU6
Gene Alias AI467640; AW121680; extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; Kiaa1077; mKIAA1077; mSulf-1; RSulfFP1; Sulf1; SULF-1; sulfatase 1; sulfatase FP
Gene Symbols SULF1
Host Species Rabbit
Immunogen Recombinant Human Extracellular sulfatase Sulf-1 protein (105-372AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23213
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.