Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119439
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119439 200 μL
PIPA5121571 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119439 Supplier Invitrogen™ Supplier No. PA5119439
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SULF2 Polyclonal Antibody for Western Blot, ICC/IF

Heparan sulfate 6-O-endosulfatases, such as Sulfatase 2, selectively remove 6-O-sulfate groups from heparan sulfate. Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SULF2
Gene Accession No. Q8IWU5
Gene Alias 2010004N24Rik; AU020235; DKFZp313E091; Extracellular sulfatase Sulf-2; Extracellular sulfatase Sulf-2-like protein; FLJ44488; FLJ90554; HSULF-2; KIAA1247; MGC126411; mKIAA1247; mSulf-2; OTTHUMP00000031223; RP5-1049G16.1; SULF2; sulfatase 2; sulfatase FP2; UNQ559/PRO1120
Gene Symbols SULF2
Host Species Rabbit
Immunogen Recombinant protein Sulfatase 2. The antigen corresponds to amino acid range 337-649 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152351, 55959
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.