Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SULF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | SULF2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119440
|
Invitrogen™
PA5119440 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121572
|
Invitrogen™
PA5121572 |
20 μL |
Each for $237.00
|
|
|||||
Description
SULF2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Heparan sulfate 6-O-endosulfatases, such as Sulfatase 2, selectively remove 6-O-sulfate groups from heparan sulfate. Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 ).Specifications
SULF2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
SULF2 | |
2010004N24Rik; AU020235; DKFZp313E091; Extracellular sulfatase Sulf-2; Extracellular sulfatase Sulf-2-like protein; FLJ44488; FLJ90554; HSULF-2; KIAA1247; MGC126411; mKIAA1247; mSulf-2; OTTHUMP00000031223; RP5-1049G16.1; SULF2; sulfatase 2; sulfatase FP2; UNQ559/PRO1120 | |
SULF2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8CFG0 | |
72043 | |
Recombinant protein Sulfatase 2. The antigen corresponds to amino acid range 337-654 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title