Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfacetamide Sodium Salt Hydrate 98.0+%, TCI America™
SDP

Catalog No. S0578250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0578250G 250 g
S057825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0578250G Supplier TCI America Supplier No. S0578250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127-56-0
Molecular Formula C8H9N2NaO3S
Molecular Weight (g/mol) 236.22
MDL Number MFCD00149555
InChI Key PQMSFAORUFMASU-UHFFFAOYSA-M
Synonym N-Acetyl-4-aminobenzenesulfonamide Sodium Salt, N-Acetylsulfanilamide Sodium Salt
PubChem CID 4022878
ChEBI CHEBI:9327
IUPAC Name sodium acetyl(4-aminobenzenesulfonyl)azanide
SMILES [Na+].CC(=O)[N-]S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Color White-Yellow
Quantity 250 g
Formula Weight 236.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfacetamide Sodium Salt Hydrate
EINECSNumber (9)-0038
RTECSNumber AC8480000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.