Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine, 98%

Catalog No. p-7082096
Click to view available options
Quantity:
250 g
1000 g

CAS: 68-35-9 | C10H10N4O2S | 250.276 g/mol

Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-35-9
Molecular Formula C10H10N4O2S
Molecular Weight (g/mol) 250.276
MDL Number MFCD00006065
InChI Key SEEPANYCNGTZFQ-UHFFFAOYSA-N
Synonym sulfadiazine, sulphadiazine, sulfapyrimidine, sulfadiazin, sulfazine, sulfadiazene, liquadiazine, sulfapyrimidin, adiazin, adiazine
PubChem CID 5215
ChEBI CHEBI:9328
IUPAC Name 4-amino-N-pyrimidin-2-ylbenzenesulfonamide
SMILES C1=CN=C(N=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 253°C to 256°C
Assay Percent Range 99%
Quantity 250 g
Merck Index 14,8903
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 250.28
Percent Purity 99%
Chemical Name or Material Sulfadiazine
Hazard Category H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 200-685-8
RTECSNumber WP1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.