Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfadimethoxine, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Specifications
| Type | Sulfadimethoxine |
| Chemical Name or Material | Sulfadimethoxine |
| Physical Form | Crystalline Powder |
| CAS | 122-11-2 |
| Melting Point | 195°C to 203°C |
| Infrared Spectrum | Conforms |
| Loss on Drying | ≤0.5 % (1 g, 105°C) |
| Packaging | Glass Bottle |
| Color | Cream to White |
| Quantity | 5 g |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction