Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadimethoxine, Thermo Scientific™

Catalog No. p-7227420
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Type Sulfadimethoxine
Chemical Name or Material Sulfadimethoxine
Physical Form Crystalline Powder
CAS 122-11-2
Melting Point 195°C to 203°C
Infrared Spectrum Conforms
Loss on Drying ≤0.5 % (1 g, 105°C)
Packaging Glass Bottle
Color Cream to White
Quantity 5 g
Molecular Formula C12 H14 N4 O4 S
Product Line Life science
MDL Number 00057345
Shipping Condition No special requirements
Molecular Weight (g/mol) 310.33
Percent Purity ≥97.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.