Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-3-iodo-L-alanine methyl ester, 97%

Catalog No. AC438460050
Click to view available options
Quantity:
1 g
5 g

CAS: 93267-04-0 | C9H16INO4 | 329.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93267-04-0
Molecular Formula C9H16INO4
Molecular Weight (g/mol) 329.13
MDL Number MFCD00216579
InChI Key UGZBFCCHLUWCQI-LURJTMIESA-N
Synonym boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
PubChem CID 10903591
SMILES COC(=O)[C@H](CI)NC(=O)OC(C)(C)C

Specifications

Melting Point 49.0°C to 52.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Specific Rotation − 3.40
IUPAC Name methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
Formula Weight 329.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material BOC-3-iodo-L-alanine methyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.