Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sulfamerazine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175430
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175430 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175430 Supplier Invitrogen™ Supplier No. PA175430
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 2.5 μg/mL. Sensitivity: 10 ng/mL Sulfamerazine produces 68% inhibition in a competitive ELISA employing Sulphamerazine polyclonal antibody.

Sulfamerazine is only found in individuals that have used or taken this drug. It is a sulfanilamide that is used as an antibacterial agent. [PubChem]Sulfamerazine is a sulfonamide drug that inhibits bacterial synthesis of dihydrofolic acid by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthetase (dihydrofolate synthetase). Sulfamerazine is bacteriostatic in nature. Inhibition of dihydrofolic acid synthesis decreases the synthesis of bacterial nucleotides and DNA. [National Center for Biotechnology Information. PubChem Compound Database; CID=5325].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfamerazine
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.3
Gene Alias C11H12N4O2S; Cremomerazine; Sulfamerazin; Sulfamethyldiazine; Sulphamerazine
Host Species Sheep
Immunogen Sulphamerazine-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.