Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethazine, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
57-68-1
C12H14N4O2S
278.33
MFCD00006066
ASWVTGNCAZCNNR-UHFFFAOYSA-N
sulfamethazine,sulfadimidine,sulfadimethyldiazine,sulfamezathine,sulfadimerazine,sulphamethazine,sulphamezathine,sulfadimezine,sulphadimidine,sulfadimesin
5327
CHEBI:102265
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C
57-68-1
C12H14N4O2S
278.33
MFCD00006066
ASWVTGNCAZCNNR-UHFFFAOYSA-N
sulfamethazine,sulfadimidine,sulfadimethyldiazine,sulfamezathine,sulfadimerazine,sulphamethazine,sulphamezathine,sulfadimezine,sulphadimidine,sulfadimesin
5327
CHEBI:102265
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C
Melting Point 197.0°C to 200.0°C
Color White to Beige
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Assay Percent Range 99%
Merck Index 15, 9047
Formula Weight 278.32
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Sulfamethazine, 99%
Show More Show Less

LIGHT SENSITIVE

EINECSNumber : 200-346-4

RTECSNumber : WO9275000

TSCA : TSCA